Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cd4
Entrez Gene ID
24932
Gene Name
Cd4 molecule
Gene Aliases
W3/25, p55
Chromosome Location
Chr.4: 157381862 - 157408358 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012705.1 NP_036837.1 3 - 4 436 69
XM_006237331.2 XP_006237393.1 5 - 6 699 69
XM_008763280.1 XP_008761502.1 3 - 4 416 69
GenBank mRNA M15768.1 - 3 - 4 436 69

View Full Product Details