Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ms4a2
Entrez Gene ID
25316
Gene Name
membrane-spanning 4-domains, subfamily A, member 2
Gene Aliases
FCIGA, Fcer1b, Ms4a1
Chromosome Location
Chr.1: 227957363 - 227965312 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012845.1 NP_036977.1 1 - 2 118 59
GenBank mRNA M22923.1 - 1 - 2 118 59
M22924.1 - 1 - 2 118 59

View Full Product Details