The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_183054.1 | NP_898877.1 | 7 - 8 | 1129 | 131 |
XM_006232631.2 | XP_006232693.1 | 7 - 8 | 131 | ||
GenBank mRNA | AY129072.1 | - | 7 - 8 | 1129 | 131 |
BC079365.1 | - | 7 - 8 | 131 |