The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017280.2 | NP_058976.1 | 8 - 9 | 579 | 99 |
GenBank mRNA | BC081817.1 | - | 8 - 9 | 579 | 99 |
D90258.1 | - | 8 - 9 | 604 | 99 | |
FQ211111.1 | - | 8 - 9 | 608 | 99 | |
FQ219582.1 | - | 8 - 9 | 699 | 99 | |
FQ224319.1 | - | - | 606 | 99 | |
FQ233787.1 | - | 8 - 9 | 641 | 99 | |
X55985.1 | - | 8 - 9 | 615 | 99 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004094.1 | NP_001004094.1 | 8 - 9 | 604 | 99 |
GenBank mRNA | M58593.1 | - | 8 - 9 | 604 | 99 |