The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031706.1 | NP_113894.1 | 3 - 4 | 235 | 104 |
GenBank mRNA | BC082802.1 | - | 2 - 3 | 219 | 104 |
BC166878.1 | - | 3 - 4 | 235 | 104 | |
FQ221315.1 | - | 3 - 4 | 235 | 104 | |
FQ222655.1 | - | 3 - 4 | 272 | 104 | |
FQ222951.1 | - | 3 - 4 | 233 | 104 | |
FQ223011.1 | - | 3 - 4 | 235 | 104 | |
FQ223286.1 | - | 3 - 4 | 237 | 104 | |
FQ229267.1 | - | 3 - 4 | 235 | 104 | |
X06423.1 | - | 3 - 4 | 235 | 104 |