Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rps8
Entrez Gene ID
65136
Gene Name
ribosomal protein S8
Gene Aliases
-
Chromosome Location
Chr.5: 136020941 - 136023511 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA FQ222655.1 - 1 - 2 62 68

View Full Product Details