Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gas2l2
Entrez Gene ID
287570
Gene Name
growth arrest-specific 2 like 2
Gene Aliases
Gas2
Chromosome Location
Chr.10: 70639429 - 70646402 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105823.2 NP_001099293.2 1 - 2 380 58

View Full Product Details