Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Htatsf1
Entrez Gene ID
317612
Gene Name
HIV-1 Tat specific factor 1
Gene Aliases
RGD1563377
Chromosome Location
Chr.X: 159514053 - 159528281 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108259.1 NP_001101729.1 3 - 4 622 110

View Full Product Details