Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nadk2
Entrez Gene ID
365699
Gene Name
NAD kinase 2, mitochondrial
Gene Aliases
Nadkd1, RGD1306809
Chromosome Location
Chr.2: 58462588 - 58504735 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044252.1 NP_001037717.1 11 - 12 1004 131
XM_006232014.2 XP_006232076.1 11 - 12 1298 131
XM_006232015.2 XP_006232077.1 10 - 11 1231 131
XM_006232016.2 XP_006232078.1 12 - 13 1084 131
GenBank mRNA BC105901.1 - 7 - 8 588 131
DQ504300.1 - 11 - 12 1004 131

View Full Product Details