The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037139.1 | NP_001032216.1 | - | 2588 | 77 |
GenBank mRNA | AY574012.1 | - | - | 2588 | 77 |