Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ncaph2
Entrez Gene ID
300149
Gene Name
non-SMC condensin II complex, subunit H2
Gene Aliases
-
Chromosome Location
Chr.7: 130326599 - 130343655 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024877.1 NP_001020048.1 19 - 20 1665 77
XM_006242186.2 XP_006242248.1 18 - 19 1630 77
XM_006242187.2 XP_006242249.1 18 - 19 1627 77
XM_006242188.2 XP_006242250.1 18 - 19 1591 77
GenBank mRNA BC097379.1 - 19 - 20 1665 77

View Full Product Details