Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Gmeb2
- Entrez Gene ID
-
83635
- Gene Name
- glucocorticoid modulatory element binding protein 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 176756006 - 176791960 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF205780.1 |
- |
- |
951 |
81 |
View Full Product Details