Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Osgep
Entrez Gene ID
290028
Gene Name
O-sialoglycoprotein endopeptidase
Gene Aliases
-
Chromosome Location
Chr.15: 27842447 - 27849866 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100510.1 NP_001093980.1 1 - 2 208 120
GenBank mRNA BC087140.1 - 1 - 2 132 120
BC158592.1 - 1 - 2 208 120

View Full Product Details