Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Osgep
Entrez Gene ID
290028
Gene Name
O-sialoglycoprotein endopeptidase
Gene Aliases
-
Chromosome Location
Chr.15: 27842447 - 27849866 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100510.1 NP_001093980.1 4 - 5 602 78
GenBank mRNA BC087140.1 - 4 - 5 526 78
BC158592.1 - 4 - 5 602 78

View Full Product Details