Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rab2b
Entrez Gene ID
305853
Gene Name
RAB2B, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.15: 28675504 - 28696192 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037645.1 NP_001032734.1 7 - 8 638 132
XM_006251883.2 XP_006251945.1 8 - 9 738 132
GenBank mRNA BC092636.1 - 2 - 3 153 132
BC107443.1 - 7 - 8 638 132

View Full Product Details