Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gpbp1l1
Entrez Gene ID
313519
Gene Name
GC-rich promoter binding protein 1-like 1
Gene Aliases
-
Chromosome Location
Chr.5: 135412256 - 135439577 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034913.1 NP_001030085.1 1 - 2 306 62
GenBank mRNA BC105913.1 - 1 - 2 306 62

View Full Product Details