Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nlgn3
Entrez Gene ID
171297
Gene Name
neuroligin 3
Gene Aliases
-
Chromosome Location
Chr.X: 71199357 - 71227460 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008773251.1 XP_008771473.1 8 - 9 4906 159

View Full Product Details