Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gata2
Entrez Gene ID
25159
Gene Name
GATA binding protein 2
Gene Aliases
-
Chromosome Location
Chr.4: 120133713 - 120142488 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033442.1 NP_254277.1 3 - 4 675 78
GenBank mRNA AB102789.1 - 1 - 2 232 78
AB102790.1 - 1 - 2 232 78
AF345897.1 - 1 - 2 232 78
AY032734.1 - 3 - 4 675 78
BC061745.1 - 2 - 3 482 78
FQ212304.1 - - 556 78

View Full Product Details