The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100750.1 | NP_001094220.1 | 1 - 2 | 89 | 71 |
GenBank mRNA | BC079367.1 | - | 1 - 2 | 82 | 71 |
BC099746.1 | - | 1 - 2 | 73 | 71 | |
BC160886.1 | - | 1 - 2 | 89 | 71 |