Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dnajb1
Entrez Gene ID
361384
Gene Name
DnaJ (Hsp40) homolog, subfamily B, member 1
Gene Aliases
-
Chromosome Location
Chr.19: 24747178 - 24750919 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108441.1 NP_001101911.1 3 - 4 974 67

View Full Product Details