The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108725.1 | NP_001102195.1 | 4 - 5 | 484 | 93 |
XM_006240639.2 | XP_006240701.2 | 4 - 5 | 540 | 93 | |
XM_006240641.2 | XP_006240703.2 | 4 - 5 | 540 | 93 | |
XM_006240642.2 | XP_006240704.2 | 4 - 5 | 540 | 93 | |
XM_006240643.2 | XP_006240705.2 | 4 - 5 | 540 | 93 | |
XM_006240644.2 | XP_006240706.2 | 4 - 5 | 540 | 93 | |
XM_008764946.1 | XP_008763168.1 | 4 - 5 | 540 | 93 |