Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Rbmxl1
- Entrez Gene ID
-
680944
- Gene Name
- RNA binding motif protein, X-linked-like 1
- Gene Aliases
-
Rbmxrt
- Chromosome Location
-
Chr.4: 42186598 - 42188294 on Build Rnor_6.0
- Gene Symbol
- Rbmxrtl
- Entrez Gene ID
-
307779
- Gene Name
- RNA binding motif protein, X chromosome retrogene-like
- Gene Aliases
-
Rbmx, Rbmxrt
- Chromosome Location
-
Chr.19: 33081473 - 33084988 on Build Rnor_6.0
View Full Product Details