The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008366.1 | NP_001008367.1 | 11 - 12 | 1157 | 71 |
XM_006255686.2 | XP_006255748.1 | 11 - 12 | 1271 | 71 | |
XM_006255687.2 | XP_006255749.1 | 11 - 12 | 1189 | 71 | |
XM_006255689.2 | XP_006255751.1 | 12 - 13 | 1493 | 71 | |
XM_006255690.2 | XP_006255752.1 | 12 - 13 | 1279 | 71 | |
XM_006255691.2 | XP_006255753.1 | 10 - 11 | 1213 | 71 | |
XM_006255692.2 | XP_006255754.1 | 10 - 11 | 1093 | 71 | |
GenBank mRNA | AY325257.1 | - | - | 1444 | 71 |
AY325261.1 | - | - | 1444 | 71 | |
BC085839.1 | - | 11 - 12 | 1157 | 71 |