Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdh15
Entrez Gene ID
361432
Gene Name
cadherin 15
Gene Aliases
-
Chromosome Location
Chr.19: 55669661 - 55689986 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207613.1 NP_997496.1 1 - 2 57 95
GenBank mRNA AB176538.1 - 1 - 2 57 95

View Full Product Details