Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ranbp2
Entrez Gene ID
294429
Gene Name
RAN binding protein 2
Gene Aliases
RGD1560047
Chromosome Location
Chr.20: 28027054 - 28076664 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191604.1 NP_001178533.1 - 213 118

View Full Product Details