Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cct4
Entrez Gene ID
29374
Gene Name
chaperonin containing Tcp1, subunit 4 (delta)
Gene Aliases
-
Chromosome Location
Chr.14: 107767392 - 107780270 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182814.2 NP_877966.1 5 - 6 659 100
GenBank mRNA AY223861.1 - 5 - 6 525 100
BC079283.1 - 5 - 6 630 100

View Full Product Details