Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc25a25
Entrez Gene ID
246771
Gene Name
solute carrier family 25 (mitochondrial carrier, phosphate carrier), member 25
Gene Aliases
Mcsc, Pcscl
Chromosome Location
Chr.3: 11442396 - 11476186 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145677.1 NP_663710.1 6 - 7 899 69
XM_006233931.2 XP_006233993.1 7 - 8 1056 69
XM_006233932.2 XP_006233994.1 6 - 7 1020 69
XM_006233933.2 XP_006233995.1 7 - 8 1708 69
XM_006233934.2 XP_006233996.1 6 - 7 1672 69
XM_006233935.2 XP_006233997.1 7 - 8 2272 69
XM_006233937.2 XP_006233999.1 7 - 8 1292 69
GenBank mRNA AY043169.1 - 6 - 7 899 69
FQ211394.1 - 6 - 7 979 69

View Full Product Details