Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc25a25
Entrez Gene ID
246771
Gene Name
solute carrier family 25 (mitochondrial carrier, phosphate carrier), member 25
Gene Aliases
Mcsc, Pcscl
Chromosome Location
Chr.3: 11442396 - 11476186 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145677.1 NP_663710.1 8 - 9 1168 81
XM_006233931.2 XP_006233993.1 9 - 10 1325 81
XM_006233932.2 XP_006233994.1 8 - 9 1289 81
XM_006233933.2 XP_006233995.1 9 - 10 1977 81
XM_006233934.2 XP_006233996.1 8 - 9 1941 81
XM_006233935.2 XP_006233997.1 9 - 10 2541 81
XM_006233937.2 XP_006233999.1 9 - 10 1561 81
GenBank mRNA AY043169.1 - 8 - 9 1168 81
FQ211394.1 - 8 - 9 1248 81

View Full Product Details