Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc25a25
Entrez Gene ID
246771
Gene Name
solute carrier family 25 (mitochondrial carrier, phosphate carrier), member 25
Gene Aliases
Mcsc, Pcscl
Chromosome Location
Chr.3: 11442396 - 11476186 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145677.1 NP_663710.1 1 - 2 256 63
XM_006233935.2 XP_006233997.1 1 - 2 1593 63
GenBank mRNA AY043169.1 - 1 - 2 256 63
FQ211394.1 - 1 - 2 272 63

View Full Product Details