Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spaca1
Entrez Gene ID
500432
Gene Name
sperm acrosome associated 1
Gene Aliases
RGD1560284
Chromosome Location
Chr.5: 49424064 - 49439180 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191982.1 NP_001178911.1 1 - 2 223 82

View Full Product Details