Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gle1
Entrez Gene ID
362098
Gene Name
GLE1 RNA export mediator
Gene Aliases
Gle1l
Chromosome Location
Chr.3: 8498086 - 8530212 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025731.1 NP_001020902.1 6 - 7 959 73
XM_006233836.2 XP_006233898.1 6 - 7 995 73
XM_006233838.2 XP_006233900.1 6 - 7 994 73
GenBank mRNA BC099088.1 - 6 - 7 959 73
FQ217346.1 - 6 - 7 980 73

View Full Product Details