The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014178.2 | NP_001014200.2 | 2 - 3 | 280 | 58 |
GenBank mRNA | AI070071.1 | - | 2 - 3 | 280 | 58 |