Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Il11ra1
Entrez Gene ID
245983
Gene Name
interleukin 11 receptor, alpha chain 1
Gene Aliases
-
Chromosome Location
Chr.5: 58149139 - 58159070 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139116.1 NP_620816.1 10 - 11 1133 122
XM_006238021.2 XP_006238083.1 9 - 10 1851 122
XM_006238022.2 XP_006238084.1 9 - 10 1271 122
XM_006238023.2 XP_006238085.1 10 - 11 1753 122
GenBank mRNA AF347936.1 - 10 - 11 1133 122
BC070921.1 - 10 - 11 1128 122

View Full Product Details