Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Qrsl1
Entrez Gene ID
309911
Gene Name
glutaminyl-tRNA synthase (glutamine-hydrolyzing)-like 1
Gene Aliases
-
Chromosome Location
Chr.20: 48855839 - 48881110 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014034.1 NP_001014056.1 5 - 6 616 92
XM_006256588.2 XP_006256650.1 5 - 6 655 92
GenBank mRNA BC089213.1 - 5 - 6 616 92

View Full Product Details