Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zbtb24
Entrez Gene ID
365590
Gene Name
zinc finger and BTB domain containing 24
Gene Aliases
Bif1a, ZNF450
Chromosome Location
Chr.20: 46166646 - 46191172 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098667.1 NP_001092137.1 4 - 5 1385 144
XM_008773001.1 XP_008771223.1 4 - 5 2915 144
XM_008773002.1 XP_008771224.1 4 - 5 1378 144

View Full Product Details