Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lca5
Entrez Gene ID
300866
Gene Name
Leber congenital amaurosis 5
Gene Aliases
RGD1308555
Chromosome Location
Chr.8: 90926309 - 90984284 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013954.1 NP_001013976.1 5 - 6 1212 117
XM_008766249.1 XP_008764471.1 4 - 5 1156 117
XM_008766250.1 XP_008764472.1 3 - 4 1037 117
XM_008766251.1 XP_008764473.1 5 - 6 1247 117
XM_008766252.1 XP_008764474.1 4 - 5 1100 117
XM_008766253.1 XP_008764475.1 4 - 5 1543 117
XM_008766255.1 XP_008764477.1 5 - 6 1272 117
XM_008766256.1 XP_008764478.1 4 - 5 1078 117
XM_008766257.1 XP_008764479.1 4 - 5 1717 117
XM_008766258.1 XP_008764480.1 4 - 5 4741 117
XM_008766259.1 XP_008764481.1 4 - 5 1157 117
GenBank mRNA BC083657.1 - 3 - 4 1033 117
BC085805.1 - 5 - 6 1212 117

View Full Product Details