The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106860.1 | NP_001100330.1 | 2 - 3 | 198 | 63 |
XM_008766512.1 | XP_008764734.1 | 3 - 4 | 363 | 63 | |
GenBank mRNA | BC166531.1 | - | 2 - 3 | 99 | 63 |