Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Yipf2
Entrez Gene ID
363027
Gene Name
Yip1 domain family, member 2
Gene Aliases
RGD1307512
Chromosome Location
Chr.8: 22621758 - 22625959 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014208.1 NP_001014230.1 7 - 8 757 88
GenBank mRNA BC083587.1 - 7 - 8 757 88

View Full Product Details