Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdsub1
Entrez Gene ID
362137
Gene Name
WD repeat, sterile alpha motif and U-box domain containing 1
Gene Aliases
-
Chromosome Location
Chr.3: 45924979 - 45955455 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014179.1 NP_001014201.1 2 - 3 602 79
XM_006234208.2 XP_006234270.1 2 - 3 862 79
XM_006234209.2 XP_006234271.1 2 - 3 861 79
XM_006234211.2 XP_006234273.1 2 - 3 583 79
GenBank mRNA BC089856.1 - 2 - 3 602 79

View Full Product Details