Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ncoa4
Entrez Gene ID
619385
Gene Name
nuclear receptor coactivator 4
Gene Aliases
-
Chromosome Location
Chr.16: 8302950 - 8323294 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034007.1 NP_001029179.1 8 - 9 1779 133
NM_001034008.1 NP_001029180.1 8 - 9 1825 133
XM_006252660.2 XP_006252722.1 8 - 9 2126 133
GenBank mRNA BC160817.1 - 8 - 9 1804 133

View Full Product Details