Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ccdc47
Entrez Gene ID
303606
Gene Name
coiled-coil domain containing 47
Gene Aliases
RGD1308813
Chromosome Location
Chr.10: 94388425 - 94406949 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013974.1 NP_001013996.1 10 - 11 1275 74
GenBank mRNA BC085824.1 - 10 - 11 1275 74

View Full Product Details