Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
A1i3
Entrez Gene ID
297568
Gene Name
alpha-1-inhibitor III
Gene Aliases
-
Chromosome Location
Chr.4: 154505735 - 154565099 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037975.2 NP_001033064.2 16 - 17 2060 150
XM_006237281.2 XP_006237343.1 16 - 17 2072 150
GenBank mRNA J03552.1 - 16 - 17 2060 150

View Full Product Details