Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tulp3
Entrez Gene ID
688749
Gene Name
tubby-like protein 3
Gene Aliases
-
Chromosome Location
Chr.4: 161636084 - 161677806 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008763338.1 XP_008761560.1 4 - 5 498 66

View Full Product Details