Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wdr81
Entrez Gene ID
303312
Gene Name
WD repeat domain 81
Gene Aliases
RGD1311334
Chromosome Location
Chr.10: 62273817 - 62287207 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134360.1 NP_001127832.1 2 - 3 3962 90

View Full Product Details