Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ttc14
Entrez Gene ID
310314
Gene Name
tetratricopeptide repeat domain 14
Gene Aliases
-
Chromosome Location
Chr.2: 120266883 - 120285472 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107666.2 NP_001101136.2 1 - 2 216 134
XM_006232257.2 XP_006232319.1 1 - 2 266 134

View Full Product Details