Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Acrbp
Entrez Gene ID
500316
Gene Name
acrosin binding protein
Gene Aliases
-
Chromosome Location
Chr.4: 157563450 - 157577045 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006237379.2 XP_006237441.1 6 - 7 1705 69

View Full Product Details