Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tsr1
Entrez Gene ID
100360406
Gene Name
TSR1, 20S rRNA accumulation, homolog (S. cerevisiae)
Gene Aliases
-
Chromosome Location
Chr.10: 61746070 - 61757518 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002727774.2 XP_002727820.1 1 - 2 129 82

View Full Product Details