Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Amdhd2
Entrez Gene ID
302972
Gene Name
amidohydrolase domain containing 2
Gene Aliases
RGD1304601
Chromosome Location
Chr.10: 13528400 - 13536990 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024990.2 NP_001020161.2 8 - 9 1041 90
XM_008767565.1 XP_008765787.1 8 - 9 1132 90
GenBank mRNA BC091278.1 - 7 - 8 1101 90

View Full Product Details