Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hs1bp3
Entrez Gene ID
313950
Gene Name
HCLS1 binding protein 3
Gene Aliases
RGD1311331
Chromosome Location
Chr.6: 33517729 - 33548015 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_233975.8 XP_233975.5 6 - 7 969 91
GenBank mRNA FQ224595.1 - 6 - 7 938 91

View Full Product Details