Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmem8a
Entrez Gene ID
303004
Gene Name
transmembrane protein 8A
Gene Aliases
M83, Tmem8
Chromosome Location
Chr.10: 15484951 - 15495380 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106991.1 NP_001100461.1 2 - 3 425 68
XM_006245973.2 XP_006246035.1 2 - 3 1039 68
XM_008767577.1 XP_008765799.1 2 - 3 2117 68

View Full Product Details